Learn More
Invitrogen™ Alpha-Synuclein [pS129] Human ProcartaPlex™ Simplex Kit
The Human Alpha-Synuclein [pS129] ProcartaPlex Kit measures Alpha-Synuclein [pS129] protein and is designed to be combinable with other Simplex kits so that users can create their own multiplex panel that utilizes Luminex xMAP technology for protein detection/quantitation.
Brand: Invitrogen™ EPX010-12361-901
Additional Details : Weight : 0.20000kg
Description
- Kit measures Alpha-Synuclein [pS129] protein
- Designed to be combinable with other Simplex kits\
- Users can create their own multiplex panel that utilizes Luminex xMAP technology for protein detection/quantitation
- When combining multiple Simplex kits (i.e., when not using a pre-configured Multiplex Panel), only one buffer kit (sold separately) is needed for each assay plate regardless of plex size
Specifications
Simplex Kit | |
Human | |
Alpha-synuclein | |
See Certificate of Analysis | |
Luminex Instruments | |
NACP, PARK1 | |
SNCA | |
<15% | |
Synuclein | |
Serum, Plasma: 25 μL; CCS: 50 μL | |
• 1 vial Capture Beads (50X) • 1 vial biotinylated Detection Antibody (50X) • 2 vials NHP Standard Mix A (lyophilized) Store at 2-8°C. |
|
Neurobiology |
96 Tests | |
Combinable | |
Alpha-Synuclein [pS129][39] | |
Simplex Kit | |
SNCA | |
6622 | |
<15% | |
ProcartaPlex™ | |
Serum, Plasma, Cell Culture Supernatants | |
P37840 | |
Wet ice |