Learn More
alpha-Toluenesulfonyl chloride, 99%, Thermo Scientific Chemicals
£17.90 - £166.00
Chemical Identifiers
CAS | 1939-99-7 |
---|---|
Molecular Formula | C7H7ClO2S |
Molecular Weight (g/mol) | 190.641 |
MDL Number | MFCD00007455 |
InChI Key | OAHKWDDSKCRNFE-UHFFFAOYSA-N |
Synonym | alpha-toluenesulfonyl chloride, benzylsulfonyl chloride, benzenemethanesulfonyl chloride, benzylsulfochloride, .alpha.-toluenesulfonyl chloride, methanesulfonyl chloride, phenyl, alpha-toluenesulfonylchloride, phenylmethylsulfonyl chloride, phenylmethanesulphonyl chloride, phenyl-methanesulfonyl chloride |
PubChem CID | 74740 |
IUPAC Name | phenylmethanesulfonyl chloride |
SMILES | C1=CC=C(C=C1)CS(=O)(=O)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11449372
|
Thermo Scientific Alfa Aesar
A10124.06 |
5 g |
£17.90
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11459372
|
Thermo Scientific Alfa Aesar
A10124.14 |
25 g |
£48.50
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11469372
|
Thermo Scientific Alfa Aesar
A10124.22 |
100 g |
£166.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1939-99-7 | |
190.641 | |
OAHKWDDSKCRNFE-UHFFFAOYSA-N | |
74740 | |
C1=CC=C(C=C1)CS(=O)(=O)Cl |
C7H7ClO2S | |
MFCD00007455 | |
alpha-toluenesulfonyl chloride, benzylsulfonyl chloride, benzenemethanesulfonyl chloride, benzylsulfochloride, .alpha.-toluenesulfonyl chloride, methanesulfonyl chloride, phenyl, alpha-toluenesulfonylchloride, phenylmethylsulfonyl chloride, phenylmethanesulphonyl chloride, phenyl-methanesulfonyl chloride | |
phenylmethanesulfonyl chloride |
Specifications
1939-99-7 | |
120°C (10 mmHg) | |
MFCD00007455 | |
972806 | |
alpha-toluenesulfonyl chloride, benzylsulfonyl chloride, benzenemethanesulfonyl chloride, benzylsulfochloride, .alpha.-toluenesulfonyl chloride, methanesulfonyl chloride, phenyl, alpha-toluenesulfonylchloride, phenylmethylsulfonyl chloride, phenylmethanesulphonyl chloride, phenyl-methanesulfonyl chloride | |
C1=CC=C(C=C1)CS(=O)(=O)Cl | |
190.641 | |
190.65 | |
alpha-Toluenesulfonyl chloride |
89°C to 94°C | |
C7H7ClO2S | |
UN3096 | |
Moisture sensitive | |
OAHKWDDSKCRNFE-UHFFFAOYSA-N | |
phenylmethanesulfonyl chloride | |
74740 | |
99% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 217-717-1
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only