Learn More
alpha Tubulin Mouse anti-Canine, Human, Mouse, Rat, Clone: 1E4C11, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66031-1-IG-150UL
Product Code. 16887535
  • £189.00 / 150µL
Description
This antibody specifically recognizes Tubulin Alpha.
Alpha Tubulin is a part of a heterodimer consisting of alpha and beta tubulin subunits. The alpha and beta tubulins, which are each about 55 kDa MW, are homologous but not identical. Alpha-beta tubulin heterodimer is the basic building block of microtubules, and this intracellular cylindrical filamentous structure is present in almost eukaryotic cells. Microtubules serve as structural supports and lines of transport within the cell, as well as serving a key role in mitosis. Microtubules of the eukaryotic cytoskeleton perform essential and diverse functions and are composed of a heterodimer of alpha and beta tubulins. The alpha and beta tubulins represent the major components of microtubules, while gamma tubulin plays a critical role in the nucleation of microtubule assembly. There are multiple alpha and beta tubulin genes, which are highly conserved among species. The gene that encodes alpha tubulin is highly similar to the mouse and rat Tuba1 gene. Northern blotting studies have shown that the gene expression is predomitly found in morphologically differentiated neurologic cells.
Specifications
Specifications
alpha Tubulin
Monoclonal
1.73 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P05213, P68363, Q6P9V9
TUBA1B
alpha Tubulin Fusion Protein Ag18034
150 μL
Primary
Canine, Human, Mouse, Rat
Antibody
IgG2b
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
1E4C11
Unconjugated
TUBA1B
alpha Tubulin, Alpha tubulin ubiquitous, TUBA1B, tubulin, Tubulin alpha, tubulin, alpha 1b
Mouse
Protein A
RUO
10376, 22143, 500929, 608864
-20°C