Learn More
Texwipe™ AlphaLite™ II Cleanroom Floor Wipes
Cut-edge polyester single ply wipe that is ideal for floor cleaning and for applying cleaning solutions and disinfectants on floors
Brand:  Texwipe™ TX3424
Product Code. 11565395
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Large size for improved application on floor surfaces
  • Size is compatible with most mopping systems
  • Improved contamination control
  • Suitable for use in Class 100, ISO 5 and Class 1000, ISO 6 environments
Specifications
Specifications
600 x 250 mm
Floor cleaning and for applying cleaning solutions and disinfectants on floors
10 Wipes/Bag, 34 Bags/Cs.