missing translation for 'onlineSavingsMsg'
Learn More

Texwipe™ AlphaSat™ CleanPak™ Presaturated Polyester Wipers

For ISO Class 3 to 5 applications

Brand:  Texwipe™ TX1039


 View more versions of this product

Product Code. 11595395

  • £288.00 / Pack of 50

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Cleanroom polyester knit wipers presaturated with various concentrations of semiconductor-grade isopropanol IPA (100% IPA wiper is designed for semiconductor tool chambers)
  • Vectra™ processed for consistent quality, low extractable ions and volatile residues, and hands-free quality inspection
  • Remove fingerprints and other contaminants from medical surfaces prior to packaging and terminal sterilization
  • Superior strength resists abrasion under rigorous use
  • Optimum IPA saturation level
  • Eliminates maintenance and safety concerns of squirt bottles
  • Lowest particulate generation when introduced to alcohol
  • Lot-to-lot traceability

Specifications

Specifications

Pre-saturated Wipe
22.9 cm
22.9 cm
158
9 x 9 in. (22.86 x 22.86 cm)
70% Semiconductor grade Isopropyl Alcohol
9 in.
9 in.
Polyester
ISO 9001:2000, 13485:2003, 14001:2004 and OHSAS 18001:1999
Ideal for general wiping, spill control, cleaning and polishing Stainless Steel, protective covering for parts and product
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Texwipe Wipers pre-wetted with 60% - 70 % IPA

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable solid Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H228-Flammable solid.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : isopropanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers