Learn More
Ansell AlphaTec™ 1500 PLUS FR - Model 111
301.12 GBP valid until 2024-12-31
Use promo code "22917" to get your promotional price.
Use promo code "22917" to get your promotional price.
Click to view available options
Size: Small, Style: W17S111
Description
- Flame retardant and anti-static SMMS nonwoven provides a good barrier to particulates and low hazard liquid sprays or splashes
- Air and water vapour permeable (breathable) to help reduce the risk of heat stress
- Optimized body fit ensures full freedom of movement when worn over heat and flame protective clothing (EN ISO 14116 Index 2 or above)
- Anti-static
Specifications
Size | Small |
Material | FR Anti-static SMS Fabric |
Color | White, Red |
Seam Type | Stitched |
Certifications/Compliance (Europe) | EN 374:2003, EN ISO 13982-1:2004 + A1:2010, EN 13034:2005 + A1:2009, EN 1073-2:2002, EN 1149-5:2008, Limited Flame Spread EN ISO 14116:2015-1 Index 1 |
Weight (Metric) | 54 gsm |
Style | W17S111 |