Learn More
Ansell AlphaTec™ 1500 PLUS Lab Coats, Model 208 Product Code.: 17111005
Add to basket
Ansell AlphaTec™ 1500 PLUS Lab Coats, Model 208
Product Code. 17111005
Medium
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
Product Code. 17111005
Brand: Ansell WH15B0020803
Add to basket
Add to basket
Short length lab coat with collar, elasticated cuffs, studded front fastening, left breast and right thigh pockets.
  • Air and water vapor permeable (breathable)
  • Silicone Free
  • Optimized body fit helps improve wearer comfort and safety
TRUSTED_SUSTAINABILITY
Specifications
Product Type General Purpose Lab Coat
Size Medium
Gender Unisex
Material SMS
Color White
Disposable Reusable
Sterility Non-sterile
Model W15B208
Seam Type Bound
Certifications/Compliance (Europe) EN ISO 13688:2013, Partial Body Protection Only, EN 13034:2005 + A1:2009
Weight (Metric) 48 gsm
Product Title
Ansell AlphaTec™ 1500 PLUS Lab Coats, Model 208 >