missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec™ 1800 COMFORT Bound - Model 195
225.32 GBP valid until 2024-12-31
Use promo code "22917" to get your promotional price.
Use promo code "22917" to get your promotional price.
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Description
- 3 piece hood
- Elasticated hood, wrists, waist and ankles
- 2-way front zipper with resealable storm flap
- Finger Loops
Specifications
Size | 3X-Large |
Material | Microporous Non-woven Polypropylene and Polyethylene Laminate |
Color | White, Blue |
Seam Type | Bound |
Coating | Anti-Static Treatment (Back, Underarm and Hood) |
Certifications/Compliance (Europe) | EN 374:2003, EN ISO 13982-1:2004 + A1:2010, EN 13034:2005 + A1:2009, EN 1073-2:2002, EN 1149-5:2008, US Ergo Certification |
For Use With (Application) | Clean Room Cleaning and Preparing, Body Painting Including Surface Inspection, Production Line Support & Maintenance, Cleaning of Plant and Machines, Maintenance of Plant and Machines, Fibre Glass Resin Applications, Handling of Powders, General Purpose Auto Aftermarket |
Style | W18B195 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'