missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec™ 1800 Ts PLUS
385.51 GBP valid until 2024-12-31
Use promo code "22917" to get your promotional price.
Use promo code "22917" to get your promotional price.
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Description
- 3 piece hood
- Elasticated hood, wrists, waist and ankles
- 2-way front zipper with resealable storm flap
- Finger Loops
- Chinstrap
Specifications
Size | X-Large |
Material | Microporous Non-woven Polyethylene Laminate |
Color | White |
Seam Type | Serged, Taped |
Certifications/Compliance (Europe) | EN 374:2003, EN 14605:2005 + A1:2009,EN ISO 13982-1:2004 + A1:2010,EN 13034:2005 + A1:2009,EN 1073-2:2002,EN 1149-5:2008,EN 14126:2003 |
Includes | Hood and single zipper with re-sealable storm flap |
For Use With (Application) | Clean Room Cleaning and Preparing, Body Painting Including Surface Inspection, Production Line Support & Maintenance, Cleaning of Plant and Machines, Maintenance of Plant and Machines, Fibre Glass Resin Applications, Handling of Powders |
Weight (Metric) | 53 gsm |
Style | W18T111 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'