missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec™ 2000 Lab Coats, Model 209
272.63 GBP valid until 2024-12-31
Use promo code "22917" to get your promotional price.
Use promo code "22917" to get your promotional price.
Click to view available options
Size:
Small
3X-Large
Description
- Proven protective barrier to low concentration liquid chemicals, liquid and particulate biological hazards
- Comfortable to wear featuring moisture vapor permeable fabric (breathable) to help reduce the risk of heat stress
- Optimized fit, improving wearer comfort and safety
- Low Linting
Specifications
Product Type | Chemical Resistant Lab Coat |
Size | 3X-Large |
Gender | Unisex |
Color | White |
Closure Type | Zipper |
Disposable | Limited-use |
Sterility | Non-sterile |
Resistance | Chemicals |
Certifications/Compliance | CE Category III, EN 14126 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'