missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec™ 2000 Standard Hood Bound - Model 503
Click to view available options
Style: W20B503
Description
- Proven protective barrier to low concentration liquid chemicals, liquid & particulate biological hazards
- Optimized head fit, improving wearer comfort and safety
- Low Linting

Specifications
Specifications
Material | Microporous Polyethylene Laminate (Non-woven) |
Color | White |
Seam Type | Bound |
Certifications/Compliance (Europe) | EN 13034:2005 + A1:2009, EN 14126:2003, EN ISO 13688:2013, Partial Body Protection Only |
Weight (Metric) | 63 gsm |
For Use With (Application) | Clean Room Cleaning and Preparing, Production Line Support & Maintenance, Maintenance of Plant and Machines, Veterinary Services |
Style | W20B503 |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ansell AlphaTec™ 2000 Standard Hood Bound - Model 503 >
Spot an opportunity for improvement?Share a Content Correction