missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec™ 2000 STANDARD Overboots Bound
598.51 GBP valid until 2024-12-31
Use promo code "22917" to get your promotional price.
Use promo code "22917" to get your promotional price.
Click to view available options
Style:
W20B406
W20B407
Description
- Proven protective barrier to low concentration liquid chemicals, liquid and particulate biological hazards
- Optimized fit, improving wearer comfort and safety
- Low Linting
Spécification
Material | Microporous Polyethylene Laminate Non-woven |
Color | White |
Seam Type | Bound |
Style | W20B407 |
For Use With (Application) | Clean Room Cleaning and Preparing, Production Line Support & Maintenance, Maintenance of Plant and Machines, Veterinary Services |
Certifications/Compliance (Europe) | EN 13034:2005 + A1:2009, EN 14126:2003, EN ISO 13688:2013, Partial Body Protection Only |
Weight (Metric) | 63 gsm |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'