Learn More
Ansell AlphaTec™ 2000 STANDARD Trousers Bound Product Code.: 16884790
Estimated Shipment: 21-07-2025
to see stock.
Add to basket
Ansell AlphaTec™ 2000 STANDARD Trousers Bound
Product Code. 16884790 Shop All Ansell Products
2X-Large
Click to view available options
Size:
X-Large
2X-Large
Product Code. 16884790
Brand: Ansell WH20B0030106
Estimated Shipment: 21-07-2025
to see stock.
Add to basket
Add to basket
Comfortable trousers featuring moisture vapor permeable fabric (breathable) to help reduce the risk of heat stress.
  • Proven protective barrier to low concentration liquid chemicals, liquid and particulate biological hazards
  • Optimized fit, improving wearer comfort and safety
  • Low Linting
TRUSTED_SUSTAINABILITY
Specifications
Size 2X-Large
Material Microporous Non-woven Polyethylene Laminate
Color White
Seam Type Bound
Certifications/Compliance (Europe) Partial Body Protection Only, EN 13034:2005 + A1:2009, EN 14126:2003, EN ISO 13688:2013
Style W20B301
For Use With (Application) Clean Room Cleaning and Preparing, Production Line Support & Maintenance, Maintenance of Plant and Machines, Veterinary Services
Product Content Correction
Product Title
Ansell AlphaTec™ 2000 STANDARD Trousers Bound >