missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec™ 2000 Ts PLUS
434.56 GBP valid until 2024-12-31
Use promo code "22917" to get your promotional price.
Use promo code "22917" to get your promotional price.
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
Description
- Comfortable to wear featuring moisture vapor permeable fabric (breathable) to help reduce the risk of heat stress
- Optimized body fit, improving wearer comfort and safety
- Low linting to help reduce the risk of cross contamination in critical areas
- Silicone free, critical in spray painting applications
Specifications
Size | X-Small |
Material | Microporous Non-woven Polyethylene Laminate |
Color | White, Green |
Seam Type | Stitched, Taped |
Certifications/Compliance (Europe) | EN 374:2003, , EN 14605:2005 + A1:2009, EN ISO 13982-1:2004 + A1:2010, EN 13034:2005 + A1:2009, EN 1073-2:2002, EN 1149-5:2018, EN 14126:2003, EN ISO 13688:2013, DIN 32781:2010 |
Includes | Hood, single zipper, thumb loops, and chin strap |
For Use With (Application) | Product Inspection, Loading and Hauling, Fibre Glass/Resin Applications/Ceramic Fibres, Body Painting Including Surface Inspection, Cleaning of Plant and Machines, Maintaining Lab Furniture and Equipment |
Weight (Metric) | 63 gsm |
Style | W20T111 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'