missing translation for 'onlineSavingsMsg'
Learn More

Ansell AlphaTec™ 23-202 Medium Weight Double-Lined PVC Gloves

Product Code. p-7215044 Shop All Ansell Products
Click to view available options
Size:
8
9
10
Unit Size:
6 pairs

Product Code. 16835190

Brand: Ansell 23202080

Only null left
Add to basket
Add to basket

Gloves are constructed with both an acrylic fleece, and 13-gauge cotton liner for low temperature applications. Rough sandblast finish enhances grip capabilities in wet, slippery conditions.

  • Rough sandblast finish enhances grip in wet conditions
  • Medium duty glove for low temperature applications
  • Double lined with acrylic fleece and 13-gauge cotton
  • Designed to protect hands at temperatures as low as -30°C
  • Compliant with FDA & EU food handling requirements
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance (Europe) EN 420:2003 + A1:2009, EN 511:2006, REACH Compliant
Applications Industrial
Color Blue
Cut Resistance Rating ANSI A2
Exterior Finish Fully Textured
Latex Free Yes
Material PVC
Product Type Chemical-resistant Gloves
Size 8
EN388 Standard 4231C
EN ISO 374-1 Type B
EN ISO 374-1 CHEMICAL LETTER CODES KLT
EN ISO 374 - 5 Yes
Glove Cuff Style Safety
Liner Material Acrylic Fleece and 100% Cotton
Series 23-202
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.