missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec™ 2500 PLUS
629.70 GBP valid until 2024-12-31
Use promo code "22917" to get your promotional price.
Use promo code "22917" to get your promotional price.
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Description
- Air and water vapour permeable (breathable) fabric helps reduce the risk of heat stress
- Ultra low linting to help reduce the risk of cross contamination in critical areas
- Achieves the highest classifications for protection from biological agents in accordance with EN 14126 and ASTM F 1671 for penetration of blood, body fluids and blood-borne pathogens
- Type 3-B liquid tight certified
Specifications
Size | Small |
Material | Microporous Non-woven Polypropylene Laminate |
Color | White |
Seam Type | Stitched, Taped |
Certifications/Compliance (Europe) | EN 374:2003, EN 14605:2005 + A1:2009, EN 14605:2005 + A1:2009, EN ISO 13982-1:2004 + A1:2010, EN 13034:2005 + A1:2009, EN 1073-2:2002, EN 1149-5:2008, EN 14126:2003 |
For Use With (Application) | Transferring Liquids and Solids, Unexpected Leakages Spills or Other Releases, Plant Disinfection and Sanitization, Tank Cleaning, Emergency Response, Spill or Leakage Cleanup, Maintenance, Contaminated Environments |
Weight (Metric) | 65 gsm |
Style | W25T122 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'