missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec™ 2500 STANDARD
373.04 GBP valid until 2024-12-31
Use promo code "22917" to get your promotional price.
Use promo code "22917" to get your promotional price.
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Description
- Air and water vapour permeable (breathable) fabric helps reduce the risk of heat stress
- Ultra low linting to help reduce the risk of cross contamination in critical areas
- Achieves the highest classifications for protection from biological agents in accordance with EN 14126 and ASTM F 1671 for penetration of blood, body fluids and blood-borne pathogens
- Anti-static
Specifications
Color | White |
For Use With (Application) | Transferring Liquids and Solids, Unexpected Leakages Spills or Other Releases, Plant Disinfection and Sanitization, Tank Cleaning, Emergency Response, Spill or Leakage Cleanup, Maintenance, Contaminated Environments |
Material | Microporous Non-woven Polypropylene Laminate |
Size | 3X-Large |
Seam Type | Welded |
Style | W25W122 |
Weight (Metric) | 65 gsm |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'