missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec™ 2500 STANDARD Apron
409.07 GBP valid until 2024-12-31
Use promo code "22917" to get your promotional price.
Use promo code "22917" to get your promotional price.
Click to view available options
Style: W25S213
Description
- Silicone Free
- Low Linting
Specifications
Size | Universal |
Material | Non-woven Microporous Polyethylene Laminate |
Color | White |
For Use With (Application) | Critical and Controlled Environments |
Style | W25S213 |
Seam Type | Sewn |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'