missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec™ 2500 STANDARD Overboots ESD Bound
749.40 GBP valid until 2024-12-31
Use promo code "22917" to get your promotional price.
Use promo code "22917" to get your promotional price.
Click to view available options
Style: W25B407
Description
- The fabric achieves the highest classifications for protection from biological agents in accordance with EN 14126 for penetration of blood, body fluids and blood-borne pathogens
- Unique material offering exceptional strength, liquid and particulate protection
- Low linting reduces risk of contamination in critical areas
Specifications
Size | 46 to 48 |
Material | Microporous Polypropylene Laminate Non-woven |
Color | White |
Seam Type | Bound |
Style | W25B407 |
For Use With (Application) | Transferring Liquids and Solids, Tank Cleaning, Spill or Leakage Cleanup, Maintenance |
Weight (Metric) | 65 gsm |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'