missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec™ 3000 Apron
602.54 GBP valid until 2024-12-31
Use promo code "22917" to get your promotional price.
Use promo code "22917" to get your promotional price.
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Style:
Y30W215
Description
- Lightweight and durable
Specifications
Size | Large |
Material | Multi-layer Barrier Laminate Fabric |
Color | Yellow |
For Use With (Application) | Unexpected Leakages Spills or Other Releases, Loading and Unloading Trucks and Vehicles, Loading Trucks & Vehicles, Product Inspection, Cleaning of Plant and Machines |
Weight (Metric) | 80 gsm |
Style | Y30W215 |
Seam Type | Welded |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'