missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec™ 3000 AVANT2 AIRline
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Description
- Exceptional protection, over 200 chemicals permeation tested, including chemical warfare agents
- Panoramic visor design ensures a good field of vision for the wearer.
- HEPA filter provides secondary protection from airline contamination and a SMC silencer ensures the noise inside the suit is always below 70 dB (even at maximum airflow).
- Emergency rip cord permits rapid doffing of the suit in cases of emergency or undue distress to the wearer.

Specifications
Specifications
Color | Yellow |
For Use With (Application) | Transferring Liquids and Solids Between Vessels and Tanks and Process Equipment, Opening and Draining Pumps Valves or Lines, Filling, Blending and Charging of Raw Materials, Mixing or Loading Chemicals, Cleaning of Plant and Machines, Unexpected Leakages Spills or Other Releases, Loading and Unloading Trucks and Vehicles, Product Inspection |
Material | Multi-layer Barrier Laminate Fabric |
Size | Medium |
Seam Type | Welded and Taped |
Style | Y300757D |
Weight (Metric) | 80 gsm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ansell AlphaTec™ 3000 AVANT2 AIRline >
Spot an opportunity for improvement?Share a Content Correction