missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec™ 4000
569.18 GBP valid until 2024-12-31
Use promo code "22917" to get your promotional price.
Use promo code "22917" to get your promotional price.
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
Description
- Exceptional protection, over 200 chemicals permeation tested, including chemical warfare agents
- Designed to protect, typical innovative features include dual zip systems and double cuffs
- Welded and taped seams, our highest barrier to liquids and particulates
- Enhanced comfort, knitted inner cuffs and inner textile type fabric improves wearer acceptance
- Anti-static
Specifications
Size | X-Small |
Material | Multi-layer Barrier Laminate Fabric |
Color | Green |
Seam Type | Welded, Taped |
Certifications/Compliance (Europe) | EN 374:2003, EN 14605:2005 + A1:2009, EN 14605:2005 + A1:2009, EN ISO 13982-1:2004 + A1:2010, EN 1073-2:2002, EN 1149-5:2008, EN 14126:2003, US Ergo Certification |
For Use With (Application) | Contaminated Environments, Chemical Handling, Filling, Blending and Charging of Raw Materials, Transferring Liquids and Solids, Opening and Draining Pumps Valves or Lines, Loading and Unloading of Proces Equipment, Raw Material Sample Collection, Concentration Mixing and Palletisation, Hazmat Response, Assembly and Disassembly of Large Pipes |
Weight (Metric) | 103 gsm |
Style | G40T111 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'