Learn More
Ansell AlphaTec™ 4000 Jacket Product Code.: 17190685
Estimated Shipment: 12-06-2025
to see stock.
Add to basket
Ansell AlphaTec™ 4000 Jacket
Product Code. 17190685
Large
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Product Code. 17190685
Brand: Ansell GR40T0023004
Estimated Shipment: 12-06-2025
to see stock.
Add to basket
Add to basket
A unique multi-layer barrier fabric renowned for its lightweight, yet robust textile feel and exceptional barrier to organic & inorganic chemicals.
  • Exceptional protection, over 200 chemicals permeation tested, including chemical warfare agents
  • Designed to protect
  • Silicone free
TRUSTED_SUSTAINABILITY
Specifications
Size Large
Material Multi-layer Barrier Laminate Fabric
Color Green
Seam Type Welded and Taped
Certifications/Compliance (Europe) Partial Body Protection Only, EN 14605:2005 + A1:2009, EN 14605:2005 + A1:2009, EN 14126:2003
Style G40T230
For Use With (Application) Contaminated Environments, Chemical Handling, Filling, Blending and Charging of Raw Materials, Transferring Liquids and Solids, Opening and Draining Pumps Valves or Lines, Loading and Unloading of Proces Equipment, Raw Material Sample Collection, Concentration Mixing and Palletisation, Hazmat Response, Assembly and Disassembly of Large Pipes
Weight (Metric) 103 gsm
Product Content Correction
Product Title
Ansell AlphaTec™ 4000 Jacket >