missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec™ 4000 Overboots
1205.09 GBP valid until 2024-12-31
Use promo code "22917" to get your promotional price.
Use promo code "22917" to get your promotional price.
Click to view available options
Style: G40T424
Description
- Exceptional protection, over 200 chemicals permeation tested, including chemical warfare agents
- Designed to protect
- Silicone free
Specifications
Size | One Size |
Material | Multi-layer Barrier Laminate Fabric |
Color | Green |
Seam Type | Welded and Taped |
Style | G40T424 |
For Use With (Application) | Hazmat Response, Contaminated Environments, Filling, Blending and Charging of Raw Materials, Transferring Liquids and Solids |
Product Type | Boot Covers |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'