missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec™ 4000 Trousers
536.89 GBP valid until 2024-12-31
Use promo code "22917" to get your promotional price.
Use promo code "22917" to get your promotional price.
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Description
- Exceptional protection, over 200 chemicals permeation tested, including chemical warfare agents
- Designed to protect
- Silicone free
Specifications
Size | Medium |
Material | Multi-layer Barrier Laminate Fabric |
Color | Green |
Seam Type | Welded and Taped |
Certifications/Compliance (Europe) | Partial Body Protection Only, EN 14605:2005 + A1:2009, EN 14605:2005 + A1:2009, EN 14126:2003 |
Style | G40T301 |
For Use With (Application) | Contaminated Environments, Chemical Handling, Filling, Blending and Charging of Raw Materials, Transferring Liquids and Solids, Opening and Draining Pumps Valves or Lines, Loading and Unloading of Proces Equipment, Raw Material Sample Collection, Concentration Mixing and Palletisation, Hazmat Response, Assembly and Disassembly of Large Pipes |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'