missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec™ 4000 Ultrasonically Welded and Taped Suit
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Description
- Specialized protection:The AlphaTec™ 4000 Ultrasonically Welded and Taped - Model 130 is a robust chemical protective suit, permeation-tested to withstand more than 200 hazardous chemicals, including chemical warfare agents
- Increased strength: This full body protection suit comes with taped and welded seams to grant users added strength and chemical barrier defense
- Heightened visibility: Designed with orange-colored material, it ensures better visibility which can reduce the risk of workplace injuries
- Improved practicality: It also possesses a chinstrap and a thumb loop, which are compatible with a range of respirators and hand protection products
- Optimized fit: Its elasticated two-piece hood, waist, cuffs and ankles simplify the donning and doffing process
- Certified anti-static capabilities: In addition, this anti-static suit complies with EN 1149-5
- CE Category III Type 3/4/5 protection
Specifications
Color | Orange |
Hood Type | Attached |
Material | Multi-layer Barrier Laminate Fabric |
Size | Small |
Ankle Style | Elasticated |
Closure Type | Single zip closure |
Collar Type | 3-piece hood |
Cuff Style | Elasticated |
Resistance | Chemical |
Seam Type | Ultrasonicaly welded and taped |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'