missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec™ 5000
753.26 GBP valid until 2024-12-31
Use promo code "22917" to get your promotional price.
Use promo code "22917" to get your promotional price.
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Description
- Outstanding permeation performance to numerous organic and inorganic chemicals, biological hazards and chemical warfare agents
- Exceptional barrier protection, providing >480 minutes breakthrough time against 14 of 15 chemicals listed in EN ISO 6529
- Designed to protect, rear entry suit with neoprene rubber face seal for a close fit to full face respirators
- Welded and taped seams, our highest barrier to liquids and particulates
- Designed to protect, typical innovative features include dual zip systems, double cuffs with knitted inner cuffs
- Multi-layer barrier material which is lightweight, yet strong and durable
- Anti-static
Specifications
Size | Small |
Material | Multi-layer Barrier Laminate Fabric |
Color | Orange |
Seam Type | Welded, Taped |
Certifications/Compliance (Europe) | EN 374:2003, EN 14605:2005 + A1:2009, EN 14605:2005 + A1:2009, EN ISO 13982-1:2004 + A1:2010, EN 1073-2:2002, EN 1149-5:2008, EN 14126:2003 |
For Use With (Application) | Contaminated Environments, Chemical Handling, Filling, Blending and Charging of Raw Materials, Transferring Liquids and Solids, Opening and Draining Pumps Valves or Lines, Loading and Unloading of Proces Equipment, Raw Material Sample Collection, Concentration Mixing and Palletisation, Hazmat Response, Assembly and Disassembly of Large Pipes, Unexpected Leakages Spills or Other Releases |
Weight (Metric) | 132 gsm |
Style | O50T103 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'