missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec™ 5000 APOLLO
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Description
- Outstanding permeation performance to numerous organic and inorganic chemicals, biological hazards and chemical warfare agents
- Exceptional barrier protection, providing >480 minutes breakthrough time against 14 of 15 chemicals listed in EN ISO 6529
- Rear mounted breathing apparatus (BA) pouch - universal fit with most BA
- Welded and taped seams, our highest barrier to liquids and particulates
- Anti-static

Specifications
Specifications
Color | Orange |
For Use With (Application) | Contaminated Environments, Chemical Handling, Filling, Blending and Charging of Raw Materials, Transferring Liquids and Solids, Opening and Draining Pumps Valves or Lines, Loading and Unloading of Proces Equipment, Raw Material Sample Collection, Concentration Mixing and Palletisation, Hazmat Response, Assembly and Disassembly of Large Pipes, Unexpected Leakages Spills or Other Releases |
Material | Multi-layer Barrier Laminate Fabric |
Size | Large |
Seam Type | Welded and Taped |
Style | O50T186B |
Weight (Metric) | 132 gsm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ansell AlphaTec™ 5000 APOLLO >
Spot an opportunity for improvement?Share a Content Correction