missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell™ AlphaTec™ 5000 Ultrasonically Welded and Taped
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Description
Made of highly visible innovative material is strong, durable and suitable for workers in extremely hazardous areas, including HAZMAT response teams.
- Outstanding permeation performance to numerous organic and inorganic chemicals, biological hazards and chemical warfare agents
- Exceptional barrier protection, providing andgt;480 minutes breakthrough time against 14 of 15 chemicals listed in EN ISO 6529
- Highly visible bright orange colour for improved worker safety
- Welded and taped seams, our highest barrier to liquids and particulates
- Designed to protect, typical innovative features include dual zip systems
- Multi-layer barrier material which is lightweight, yet strong and durable
- Anti-static, tested according to EN 1149-5
Specifications
Specifications
Product Type | Chemical Protective Clothing |
Size | Small |
Gender | Unisex |
Material | Multi-layer Barrier Laminate Fabric |
Color | Orange |
Disposable | Reusable |
Waist Style | Elastic |
Ankle Style | Elastic |
Cuff Style | Double, Elastic |
Neck Style | Collared |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction