Learn More
Ansell™ AlphaTec™ AquaDri™ Chemical Gloves
Product Code. 15355161
Click to view available options
Size:
10
11
Two-layer construction keeps hands dryer for longer and with greater comfort and performance. Superior nitrile film provides advanced chemical protection against incidental exposure to bases, oils, fuels, some solvents, greases and animal fats.
  • Inner soft foam coating for moisture absorption
  • Nitrile film provides advanced chemical protection
  • Reversed-lozenge finish provides improved dry and wet grip
  • Nitrile compound for snag, puncture and abration protection
  • Foldable gutter cuff to prevent dripping on forearm
TRUSTED_SUSTAINABILITY
Specifications
Color Green
Material Nitrile
Product Type Immersion Gloves
Size 11
Thickness (Metric) 0.3 mm
Glove Cuff Style Gauntlet
Liner Material Aquadri™ 2
Quantity 12 Pairs
Resistant To Chemical resistance
Series 58-330