Learn More
Ansell AlphaTec™ 58-535B Series Nitrile Immersion Gloves
Enables users to handle wet or oily objects with less grip force and more control. The patented Ansell Grip TechnologyTM ensures that the coating polymer does not penetrate the liner during manufacture significantly reducing the likelihood of chemical leakage onto the skin.
£68.70
Specifications
Certifications/Compliance | FDA 21 CFR 177-2600 |
---|---|
Glove Cuff Style | Gauntlet |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
11766703
|
Ansell
58535B110 |
11 |
£68.70
6 pairs |
Estimated Shipment: 24-04-2025
Log in to see stock. |
|||||
11776703
|
Ansell
58535B070 |
7 |
£68.70
6 pairs |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||||
11786703
|
Ansell
58535B080 |
8 |
£68.70
6 pairs |
In Stock
Log in to see stock. |
|||||
11796703
|
Ansell
58535B090 |
9 |
£68.70
6 pairs |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||||
11756703
|
Ansell
58535B100 |
10 |
£68.70
6 pairs |
In Stock
Log in to see stock. |
|||||
Description
Provides dexterity with resistance to cuts, punctures, and abrasions as well as petrochemicals, oils, greases, pesticides, and other agricultural chemicals- Chemical resistant
- Material fully coated nitrile gloves
- Thickness: 11 mil
- Embossed grip surface for superior wet and oil grip
- Red with black coating
- Comfortable design reduces fatigue
Specifications
FDA 21 CFR 177-2600 | |
Gauntlet |