missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec™ 87-085 Lightweight Food-Safe Latex Gloves
missing translation for 'orderingAttributeHoverText'
Size:
6.5
7.5
8.5
9.5
Description
- For use in low hazard areas
- Beaded cuff for tear resistance and easy donning
- Resistant, flexible, and comfortable
- 100% cotton flock lining
- AQL 0.65
Specifications
Specifications
Certifications/Compliance (Europe) | CE 0493, EN 420:2003 + A1:2009, REACH Compliant |
Color | Pink |
Hand | Specific |
Material | Latex |
Size | 6.5 |
EN388 Standard | 2010X |
EN ISO 374-1 | Type B |
EN ISO 374-1 CHEMICAL LETTER CODES | KLT |
EN ISO 374 - 5 | Yes |
Liner Material | 100% Cotton |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'