missing translation for 'onlineSavingsMsg'
Learn More

Ansell AlphaTec™ 87-108 Heavy Duty Chemical Protection Glove

Product Code. 17215106
Click to view available options
Size:
7.5
8.5
9.5
10.5

AlphaTec™ 87-108 Heavy Duty Chemical Protection Gloves feature good mechanical and chemical protection in heavy duty environments

  • High resistance to water-based chemicals
  • Good flexibility and elasticity preventing tears
  • Chlorinated to harden and cleanse the surface of the glove
  • AQL 0.65
TRUSTED_SUSTAINABILITY

Specifications

Applications Industrial
Color Black
Latex Free No
Material Rubber
Size 7.5
Style 87-108
Thickness (English) 59 mil
Thickness (Metric) 1.5 mm
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'