missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell™ AlphaTec™ Glove Connector
Click to view available options
Ansell™ Glove Connector
Description
Innovative design utilising the latest polymer technology that works with a wide variety of chemical glove thicknesses.
- Designed with a ribbed cone and collar for secure attachment.
- Innovative design utilising the latest polymer technology
- Creates a liquid tight seal between glove and cuff
- Consistent and reliable alternative to taping
- Quick and easy fit - improves productivity
- Works with a wide variety of chemical glove thicknesses
- Ribbed cone and collar for secure attachment
- AlphaTec™ Advanced Chemical Protection
Specifications
Certifications/Compliance (Europe) | Tested in accordance with ISO 17491-3:2008 - Determination of resistance to penetration by a jet of liquid (jet test) |
Color | Blue |
Product Type | Glove Connector |
Sterility | Non-sterile |
For Use With (Application) | Contaminated Environments|Chemical Handling|Filling, blending and charging of raw materials|Transferring liquids and solids|Opening and draining pumps valves or lines|Loading and unloading of proces equipment|Raw material sample collection|Concentration mixing and palletisation|Hazmat response|Assembly and disassembly of large pipes |
For Use With (Equipment) | Creates a liquid tight seal between glove and cuff |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'