missing translation for 'onlineSavingsMsg'
Learn More

Ansell AlphaTec™ 38-628 Butyl/Viton Immersion Gloves

Product Code. 17814851 Shop All Ansell Products
Click to view available options
Size:
7
8
11
Unit Size:
1 pair
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 17814851

Brand: Ansell 38628070

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

AlphaTec™ Gloves 38-628 offer a combination of butyl and viton layer of 0.7 mm for a maximum protection against hazardous chemicals. Provide the best resistance to the most aggressive chemicals without compromising dexterity or comfort. Offer superior protection against aliphatic, halogenated and aromatic hydrocarbons (benzene, toluene, xylene), as well as concentrated mineral acids.

  • AlphaTec™ 38-628 offers superior protection against aliphatic, halogenated and aromatic hydrocarbons (benzene, toluene, xylene), as well as concentrated mineral acids
  • The glove is very flexible and provides excellent dexterity
  • Designed for comfort, with a natural, curved ergonomic shape and soft feel, offering easy donning and good grip
  • The absence of latex proteins means there's no risk of latex allergies
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 2
Certifications/Compliance (Europe) CE Number, EN ISO 374-1:2016, EN 388:2003, EN 388:2016, EN 388:2016 +A1:2018, EN 420:2003 + A1:2009, EN ISO 21420:2020, EN ISO 374-1:2016, EN ISO 374-5:2016, EN 374:2003, , EN ISO 374-5:2016, Category III, REACH Compliant
Applications Industrial
Color Black
Finger Thickness (English) 27.6 mil
Finger Thickness (Metric) 0.7 mm
Latex Free Yes
Material Viton, Butyl Polymer
Palm Thickness (English) 27.6 mil
Palm Thickness (Metric) 0.7 mm
Product Type Butyl Gloves
Size 7
Thickness (English) 27.6 mil
Thickness (Metric) 0.7 mm
EN388 Standard 2120A
EN 421 No
EN ISO 374-1 Type A
EN ISO 374-1 CHEMICAL LETTER CODES ABCDFLNS
Glove Cuff Style Beaded/Rolled
Grip Design Smooth
Lining Unlined
Model AlphaTec™ 38-628
Length (English) 13.8 in.
Length (Metric) 350 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.