missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ AlphaTec™MICROGARD™2000 Model 417 Overshoes

Product Code. 15815352
Click to view available options
Size:
Small
Medium
Large
This item is not returnable. View return policy
This item is not returnable. View return policy

MICROGARD™ 2000 STANDARD provides bothprotection and comfort with exceptional liquid andparticulate protection. Ideal for a wide range ofindustrial applications

  • Elasticated opening
  • Bound seams
  • ESD PVC Sole
  • Size: 39-42 • 42-46 • 46-48
  • Color: White
TRUSTED_SUSTAINABILITY

Specifications

Size Medium
Latex Free Yes
Color White
Disposable Single-use
Sterility Non-sterile
Closure Type Elastic Opening
Description Overshoes - ESD
Seam Type Bound
Style Alphatec™ 2000 Overshoes - ESD Size 42-46
For Use With (Application) Proven barrier to low concentration liquid chemicals, diluted pesticides,liquid & particulate biological hazards
Certifications/Compliance EN14126; EN1073-2; DIN32781; EN1149-5; Type 4-B; Type5-B; Type6-B
Packaging Quantity 250/Pk.
Resistance Static, Bloodborne Pathogens, Chemical-resistant, Liquid, Water
Product Type Shoe Covers
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'