missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell™ AlphaTec™MICROGARD™2000 Model 417 Overshoes
Shop All Ansell Products
Click to view available options
Size:
Small
Medium
Large
Description
- Elasticated opening
- Bound seams
- ESD PVC Sole
- Size: 39-42 • 42-46 • 46-48
- Color: White

Specifications
Specifications
Size | Small |
Latex Free | Yes |
Color | White |
Disposable | Single-use |
Sterility | Non-sterile |
Closure Type | Elastic Opening |
Description | Overshoes - ESD |
Seam Type | Bound |
Style | Alphatec™ 2000 Overshoes - ESD Size 39-42 |
For Use With (Application) | Proven barrier to low concentration liquid chemicals, diluted pesticides,liquid & particulate biological hazards |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ansell™ AlphaTec™MICROGARD™2000 Model 417 Overshoes >
Spot an opportunity for improvement?Share a Content Correction