missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell™ AlphaTec™MICROGARD™2000 Model 417 Overshoes
564.91 GBP valid until 2024-12-31
Use promo code "22917" to get your promotional price.
Use promo code "22917" to get your promotional price.
Click to view available options
Size:
Small
Medium
Large
Description
- Elasticated opening
- Bound seams
- ESD PVC Sole
- Size: 39-42 • 42-46 • 46-48
- Color: White
Specifications
Size | Large |
Latex Free | Yes |
Color | White |
Disposable | Single-use |
Sterility | Non-sterile |
Closure Type | Elastic Opening |
Description | Overshoes - ESD |
Seam Type | Bound |
Style | Alphatec™ 2000 Overshoes - ESD Size 46-48 |
For Use With (Application) | Proven barrier to low concentration liquid chemicals, diluted pesticides,liquid & particulate biological hazards |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'