missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ ALPS5000 Automated Plate Heat Sealer
Fast, reliable and quiet automated plate sealing solution for any lab.
Brand: Thermo Scientific™ AB-5000
23278.85 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
The Thermo Scientific ALPS5000 is the next generation in plate sealing equipment offering fast sealing times, increased process customization and all electric operation improving reliability and reducing noise.
- Electrically Driven with Tool Free Loading
- No need to connect to an external air source or replace expensive vacuum cups
- Reduced risk of contamination inherent in pneumatically driven sealers
- Quieter operation when compared to pneumatically driven systems
- Tool free loading makes reloading foil simple and repeatable
- Touchscreen Control and Savable Protocols
- Scalable to meet labs needs now and into the future
- Precise control over sealing parameters to ensure repeatability
- Easily integrated into automation or robotics workflows
- Sealing of Multiple Plate Heights
- Seal plates from 8mm to 46mm in height with no adapters required
- Compatible with high throughput automated applications
Specifications
50/60 Hz | |
UL/NRTL, CE | |
19 x 9 x 21 in. | |
21 in. | |
19 in. | |
1 Ea. | |
55 lb. | |
9 in. | |
Plate Sealer |
110/240 V | |
Automated Plate Heat Sealer | |
110/240 V, 50/60 Hz | |
525 mm | |
482.6 mm | |
Ambient to 199°C | |
25 kg | |
228 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction