Learn More
Thermo Scientific™ ALPS30 Manual Heat Sealer
The Thermo ScientificTM ALPS 30 manual heat sealer is a hand operated microplate heat sealer that allows you to protect your samples from evaporation and contamination.
Brand: Thermo Scientific™ AB-0400
2422.16 GBP valid until 2024-12-31
Use promo code "23291" to get your promotional price.
Additional Details : Weight : 4.21020kg
Description
Protect your samples from evaporation and contamination when performing PCR or sample storage with the Thermo ScientificTM ALPS 30 manual heat sealer. Perfect for sealing small numbers of plates in low- to medium-throughput applications, the ALPS30 unit is small enough to keep on the bench and not take up a lot of space. Its simple, ergonomic manual operation provides quick, efficient sealing at your fingertips.
- Compatible with virtually any height microplate or block, including PCR plates
- Comes standard with three adapters to accommodate most plate types
- Can be operated with the wide range of Thermo Scientific heat seals for any application, including pierceable, optically-clear and permanent seals
- Consistent heat seal provides dependable protection for your PCR or sample storage plates
- Prevents contamination and evaporation, yielding more reliable results
- Features a built-in thermostat to prevent overheating
- LED indicator lights show when power is on and when unit is heating
- Small and lightweight–easy to carry
- Small footprint–doesn’t take up a lot of bench space
- Three adapters included to accommodate most plate types:
- PCR Plates
- Deepwell plates and blocks
- Standard-height 96- or 384-format plates
Specifications
3.15/1.6 A | |
110-120/220-230 V | |
Manual Heat Sealer | |
110/120 V, 220/230 V nominalIEC C13, 3-Core, 5 A min. | |
9.1 in. | |
9.8 in. | |
1 Ea. | |
8.8 lb. | |
8.7 in. |
50/60 Hz | |
CE marked | |
250 x 220 x 230 mm (9.8 x 8.7 x 9.1 in.) | |
Microplates | |
20 to 80%, non-condensing | |
250 mm | |
170°C (Non-adjustable) ±2° | |
4 kg | |
220 mm |