Learn More
Alternate BTEX Standard Formulation, High Level, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Brand: SPEX Certiprep BTEX-100H
Code : NEW
Additional Details : CAS Number : 108-88-3 Weight : 0.05000kg Transport : UN number : 1230 Chem class : 3 Pack group : II
Includes: Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Analyte | CAS No. | Concentration |
---|---|---|
Benzene | 71-43-2 | 2000μg/mL |
Ethylbenzene | 100-41-4 | 2000μg/mL |
o-Xylene | 95-47-6 | 2000μg/mL |
Toluene | 108-88-3 | 2000μg/mL |
m-Xylene | 108-38-3 | 1000μg/mL |
p-Xylene | 106-42-3 | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Alternate BTEX Standard Formulation (High Level) | |
1 mL | |
95-47-6,108-88-3,100-41-4,108-38-3,106-42-3, 106-42-3 | |
GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation | |
Methanol-P&T |
2mL amber prescored glass ampule | |
99.0 | |
Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity. | |
Benzene (2000μg/mL),Ethylbenzene (2000μg/mL),o-Xylene (2000μg/mL) |
Safety and Handling
- Alternate BTEX Standard Formulation (High Level)
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Carcinogenicity Category 1A
- Flammable liquid Category 2
- Germ cell mutagenicity Category 1B
- Reproductive toxicity Category 2
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H331-Toxic if inhaled.
- H340-May cause genetic defects.
- H350-May cause cancer.
- H361-Suspected of damaging fertility or the unborn child.
- H370-Causes damage to organs.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : benzene, toluene, ethylbenzene, methanol
Product intended for laboratory use only