missing translation for 'onlineSavingsMsg'
Learn More

Alumina A, Activity: Super I, MP EcoChrom™, 63 to 200 μm, MP Biomedicals™

Product Code. 11353589
Click to view available options
Quantity:
500 g
1 kg
5 kg
50 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

MP EcoChrom™ Alumina A - Super I is commonly used for purification processes of all kinds including sample preparation for pesticides environmental analysis, solvent purification and all kinds of chromatography requiring the highest precision and reproducibility.

  • Adsorption capacity approximately twice as high as that of MP EcoChrom™ Alumina N, Activity I.
  • Inital activity is kept in a very narrow range.
  • Extremely constant deactivation behavior.
  • Commonly used for purification processes requiring the highest precision and reproducibility.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
SMILES [O--].[O--].[O--].[Al+3].[Al+3]

Specifications

Color Pink to White
Physical Form Free-flowing Powder
Quantity 1 kg
IUPAC Name dialuminum;oxygen(2-)
Formula Weight 101.96
Density 3.94 g/cm3 (at 20°C)
Chemical Name or Material Alumina A, Activity: Super I

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use. For more information, please contact our customer service department.

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'