missing translation for 'onlineSavingsMsg'
Learn More

Alumina A, Activity: Super I, MP EcoChrom™, 63 to 200 μm, MP Biomedicals™
BIOPHARMA

Product Code. 11373589 Shop All MP Biomedicals Products
Click to view available options
Quantity:
500 g
1 kg
5 kg
50 kg
Unit Size:
1kg
500g
50kg
5kg
This item is not returnable. View return policy

Product Code. 11373589

Brand: MP Biomedicals 0204601

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MP EcoChrom™ Alumina A - Super I is commonly used for purification processes of all kinds including sample preparation for pesticides environmental analysis, solvent purification and all kinds of chromatography requiring the highest precision and reproducibility.

  • Adsorption capacity approximately twice as high as that of MP EcoChrom™ Alumina N, Activity I.
  • Inital activity is kept in a very narrow range.
  • Extremely constant deactivation behavior.
  • Commonly used for purification processes requiring the highest precision and reproducibility.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
SMILES [O--].[O--].[O--].[Al+3].[Al+3]

Specifications

Color Pink to White
Physical Form Free-flowing Powder
Quantity 50 kg
IUPAC Name dialuminium(3+) trioxidandiide
Formula Weight 101.96
Density 3.94 g/cm3 (at 20°C)
Chemical Name or Material Alumina A, Activity: Super I

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use. For more information, please contact our customer service department.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.