missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alumina A, Activity: Super I, MP EcoChrom™, 63 to 200 μm, MP Biomedicals™
Shop All MP Biomedicals Products

Click to view available options
Quantity:
500 g
1 kg
5 kg
50 kg
Unit Size:
1kg
500g
50kg
5kg
Description
- Adsorption capacity approximately twice as high as that of MP EcoChrom™ Alumina N, Activity I.
- Inital activity is kept in a very narrow range.
- Extremely constant deactivation behavior.
- Commonly used for purification processes requiring the highest precision and reproducibility.
Specifications
Specifications
| Color | Pink to White |
| Physical Form | Free-flowing Powder |
| Quantity | 5 kg |
| IUPAC Name | dialuminium(3+) trioxidandiide |
| Formula Weight | 101.96 |
| Density | 3.94 g/cm3 (at 20°C) |
| Chemical Name or Material | Alumina A, Activity: Super I |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use. For more information, please contact our customer service department.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction