missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alumina B, 32-63μM, Active, MP Biomedicals™
For Use in Column Chromatography and Adsorptive Filtration
Brand: MP Biomedicals 0202040
This item is not returnable.
View return policy
Description
Alumina is used as an adsorbent, desiccating agent, and catalyst, and in the manufacture of dental cements and refractories
Specifications
500 g | |
Alumina B | |
MFCD00003424 | |
1344-28-1 |
Store at Room Temperature. | |
Powder | |
3.94g/cm3 at 20°C (Lit.) |
Special Offers
-
Multiple Manufacturers
Dairy and Plant-Based Products Testing Promotion
Quality testing products for all your needs.Promo Code: 24134Expires: 2025-03-31
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Alumina B, 32-63μM, Active, MP Biomedicals™ > 500g
Spot an opportunity for improvement?Share a Content Correction