missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alumina B, Activity: Super I, Basic pH, MP EcoChrom™, MP Biomedicals™
Click to view available options
Quantity:
500 g
1 kg
5 kg
50 kg
Description
- MP EcoChrom™ Alumina - Super I is essentially aluminum oxide, Al2O3, occurring in nature as minerals bauxite, bayerite, corundum, etc.
- MP Alumina B - Super I show absorption capacity approximately twice as high as an Alumin B, Activity I.
- The initial activity is kept in a very narrow range and their deactivation behaviour is extremely constant.
- The product is available as a neutral version, MP Alumina N - Super I, and an acidic version, MP Alumina A - Super I.
- MP EcoChro Alumina B - Super I is commonly used for purification processes of all kinds including sample preparation for pesticides, environmental analysis, solvent purification and all kinds of chromatography requiring the highest precision and reproducibility.
Specifications
Specifications
Color | Pink to White |
Physical Form | Free-flowing Powder |
Quantity | 5 kg |
IUPAC Name | dialuminum;oxygen(2-) |
Formula Weight | 101.96 |
Density | 3.94 g/cm3 (at 20°C) |
Chemical Name or Material | Alumina B, Activity: Super I |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use. For more information, please contact our customer service department.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'