missing translation for 'onlineSavingsMsg'
Learn More

Alumina B, Activity: Super I, Basic pH, MP EcoChrom™, MP Biomedicals™

Product Code. 11393579
Click to view available options
Quantity:
500 g
1 kg
5 kg
50 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

MP EcoChrom™ Alumina is essentially aluminum oxide, Al2O3, occurring in nature as minerals bauxite, bayerite, corundum, etc. This Alumina is produced at the highest activity available in the market.

  • MP EcoChrom™ Alumina - Super I is essentially aluminum oxide, Al2O3, occurring in nature as minerals bauxite, bayerite, corundum, etc.
  • MP Alumina B - Super I show absorption capacity approximately twice as high as an Alumin B, Activity I.
  • The initial activity is kept in a very narrow range and their deactivation behaviour is extremely constant.
  • The product is available as a neutral version, MP Alumina N - Super I, and an acidic version, MP Alumina A - Super I.
  • MP EcoChro Alumina B - Super I is commonly used for purification processes of all kinds including sample preparation for pesticides, environmental analysis, solvent purification and all kinds of chromatography requiring the highest precision and reproducibility.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
SMILES [O--].[O--].[O--].[Al+3].[Al+3]

Specifications

Color Pink to White
Physical Form Free-flowing Powder
Quantity 50 kg
IUPAC Name dialuminum;oxygen(2-)
Formula Weight 101.96
Density 3.94 g/cm3 (at 20°C)
Chemical Name or Material Alumina B, Activity: Super I

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use. For more information, please contact our customer service department.

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'