missing translation for 'onlineSavingsMsg'
Learn More

Aluminum ammonium sulfate dodecahydrate, 99%, pure, Thermo Scientific Chemicals

£9.70 - £423.94

Chemical Identifiers

CAS 7784-26-1
MDL Number MFCD00149958
InChI Key WZUKKIPWIPZMAS-UHFFFAOYSA-K
Synonym ammonia alum, unii-5c36drl9zn, aluminum ammonium sulfate dodecahydrate, aluminum ammonium disulfate dodecahydrate, ammonium aluminum sulfate hydrate, alum, ammonium usp, aluminum ammonium sulfate dodecahydrate, ammonium aluminum sulfate dodecahydrate, 5c36drl9zn, ammonium aluminum disulfate dodecahydrateShow MoreShow Less
PubChem CID 62668
IUPAC Name aluminum;azanium;disulfate;dodecahydrate
SMILES [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3]
View More Specs

Products 5
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10134500
SDS View Documents
Thermo Scientific Acros
206260050
5 g Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10628704
SDS View Documents
Thermo Scientific Acros
206261000
100 g Plastic bottle
£19.20
100g
Please to purchase this item. Need a web account? Register with us today!
10450481
SDS View Documents
Thermo Scientific Acros
206260010
1 kg Plastic bottle
£86.03
1kg
Please to purchase this item. Need a web account? Register with us today!
10460481
SDS View Documents
Thermo Scientific Acros
206260025
2.5 kg Plastic bottle
£171.86
2.50kg
Please to purchase this item. Need a web account? Register with us today!
11934261
SDS View Documents
Thermo Scientific Acros
206260100
10 kg Plastic drum This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Aluminum ammonium sulfate dodecahydrate, 99%, pure, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.