Learn More
Bochem™ Aluminum Anodized Lab Jacks
Manufactured according to DIN 12897 standards. Bochem™ aluminum Anodized Lab Jacks features a rounded, east to use ergonomic knob.
Brand:  Bochem™ 11016
 View more versions of this product
Product Code. 10325911
  • £174.00 / Each
Estimated Shipment: 14-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Improved scissor technology with reduced tolerances
  • Rounded, easy to use ergonomic knob
  • Scissor-construction made of 18/10-stainless steel
  • Maximal dynamic operating load (Kg max. dyn.) and the maximal static operating load (Kg max. stat.) norms are DIN 12897 compliant
  • Type 1 (240x240mm) uses a hand-operated adjusting wheel
  • Type 2 (300x300mm) includes ratchet, which connects to adjusting wheel
Specifications
Specifications
Anodized Aluminum, Stainless Steel
100 mm
Wheels
1 Ea.
100 x 100 mm
100 mm
Lab Jack