missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum chloride, 98.5%, extra pure, anhydrous, powder, Thermo Scientific Chemicals
15.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Lewis acid
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 194.0°C |
Color | Gray or White to Yellow |
Physical Form | Powder |
Quantity | 100 g |
Assay Percent Range | 98% min. (Argentometry) |
Linear Formula | AlCl3 |
Fieser | 01,24; 02,21; 03,7; 04,10; 05,10; 06,17; 07,7; 08,13; 09,11; 10,9; 11,25; 12,26; 13,15; 14,20; 15,10; 16,10; 17,15 |
Merck Index | 15, 333 |
Solubility Information | Solubility in water: reacts. Other solubilities: freely miscible with benzene, nitrobenzene,, benzophenone, carbon tetrachloride and chloroform |
Formula Weight | 133.34 |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS05_Corrosive.png)
Product Identifier
- Aluminium chloride
- Danger
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- EUH014-Reacts violently with water.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'